Merck
  • 5-Bromovinyl 2'-deoxyuridine phosphorylation by mitochondrial and cytosolic thymidine kinase (TK2 and TK1) and its use in selective measurement of TK2 activity in crude extracts.
5-Bromovinyl 2'-deoxyuridine phosphorylation by mitochondrial and cytosolic thymidine kinase (TK2 and TK1) and its use in selective measurement of TK2 activity in crude extracts.
Nucleosides, nucleotides & nucleic acids (2008-07-05)
Liya Wang, Staffan Eriksson
ABSTRACT
Mitochondrial thymidine kinase (TK2) is responsible for phosphorylation of thymidine and deoxycytidine and plays a crucial role in mitochondrial DNA precursor synthesis. TK2 is expressed in all tissues at low levels complicating accurate determinations, especially in tissues with high cytosolic thymidine kinase (TK1) activity. Recently, 5-bromovinyl 2 '-deoxyuridine (BvdU) at 0.2 micro M was used to measure TK2 activity selectively. BvdU phosphorylation by pure human TK2 and TK1 was tested here, and the ratio of BvdU phosphorylation by TK2/TK1 was 91 at 0.2 micro M but was 500 at 2.5 micro M. Therefore, for reliable measurement of TK2 activity higher BvdU concentration should be used.
Sigma-Aldrich
(E)-5-(2-Bromovinyl)-2′-deoxyuridine