MilliporeSigma
Nanohybrids for controlled antibiotic release in topical applications.
International journal of antimicrobial agents (2007-02-17)
L Tammaro, U Costantino, A Bolognese, G Sammartino, G Marenzi, A Calignano, S Tetè, F Mastrangelo, L Califano, V Vittoria
ABSTRACT
New polymeric composite materials containing a nanohybrid to be used for the controlled release of an antibiotic molecule, chloramphenicol succinate, have been formulated, prepared and characterised. The nanohybrid consists of a layered double hydroxide of Mg-Al hydrotalcite-type, in which the nitrate anions present in the host galleries were replaced with chloramphenicol succinate anions (CFS(-)) by a simple ion-exchange reaction. Different amounts of the hybrid material were incorporated in polycaprolactone and processed as films of 0.15mm thickness. The composite materials were analysed by X-ray diffractometry and thermogravimetry and their mechanical properties were determined. They showed properties even better than those of the pristine polymer. The release process of the antibiotic molecules was found to be very interesting and promising for tuneable drug delivery. It consists of two stages: an initial stage of a very rapid burst, in which a small fraction of drug is released; and a second stage that is much slower, extending for a longer and longer time. This behaviour is profoundly different and much slower than that of a sample in which the antibiotic molecule is directly incorporated into the polymeric matrix. The parameters influencing drug release have been individuated and discussed.
Product Description
Chloramphenicol sodium succinate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chloramphenicol succinate sodium salt, ≥80% (HPLC)