MilliporeSigma
  • Photocatalytic degradation of an organophosphorus pesticide phosalone in aqueous suspensions of titanium dioxide.
Photocatalytic degradation of an organophosphorus pesticide phosalone in aqueous suspensions of titanium dioxide.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2004-05-11)
N Daneshvar, M J Hejazi, B Rangarangy, A R Khataee
ABSTRACT
The present work deals with photocatalytic degradation of an organophosphorus pesticide, phosalone, in water in the presence of TiO2 particles under UV light illumination (1000 W). The influence of the basic photocatalytic parameters such as pH of the solution, amount of TiO2, irradiation time, stirring rate, and distance from UV source, on the photodegradation efficiency of phosalone was investigated. The degradation rate of phosalone was not high when the photolysis was carried out in the absence of TiO2 and it was negligible in the absence of UV light. The half-life (DT50) of a 20 ppm aqueous solution of phosalone was 15 min in optimized conditions. The plot of lnC (phosalone) vs. time was linear, suggesting first order reaction (K=0.0532 min(-1)). The half-life time of photomineralization in the concentration range of 7.5-20 ppm was 13.02 min. The efficiency of the method was also determined by measuring the reduction of Chemical Oxygen Demand (COD). During the mineralization under optimized conditions, COD decreased by more than 45% at irradiation time of 15 min. The photodegradation of phosalone was enhanced by addition of proper amount of hydrogen peroxide (150 ppm).
Product Description
Supelco
Phosalone, PESTANAL®, analytical standard