MilliporeSigma
  • The new nucleoporin: regulator of transcriptional repression and beyond.
The new nucleoporin: regulator of transcriptional repression and beyond.
Nucleus (Austin, Tex.) (2012-10-11)
Nayan J Sarma, Kristine Willis
ABSTRACT
Transcriptional regulation is a complex process that requires the integrated action of many multi-protein complexes. The way in which a living cell coordinates the action of these complexes in time and space is still poorly understood. Recent work has shown that nuclear pores, well known for their role in 3' processing and export of transcripts, also participate in the control of transcriptional initiation. We have recently begun to explore how nuclear pores interface with the well-described machinery that regulates initiation. This work led to the discovery that specific nucleoporins are required for binding of the repressor protein Mig1 to its site in target promoters. Nuclear pores are therefore involved in repressing, as well as activating, transcription. Here we discuss in detail the main models explaining our result and consider what each implies about the roles that nuclear pores play in the regulation of gene expression.
Product Description
Supelco
Invertase from baker′s yeast (S. cerevisiae), 200-300 units/mg solid
Sigma-Aldrich
Invertase Glycoprotein Standard, BioReagent, from Saccharomyces cerevisiae
Sigma-Aldrich
Invertase from baker′s yeast (S. cerevisiae), Grade VII, ≥300 units/mg solid
Sigma-Aldrich
Invertase from Candida utilis, Grade X, ≥300 units/mg solid