MilliporeSigma
  • The endoplasmic reticulum membrane complex promotes proteostasis of GABAA receptors.
The endoplasmic reticulum membrane complex promotes proteostasis of GABAA receptors.
iScience (2022-08-09)
Angela L Whittsette, Ya-Juan Wang, Ting-Wei Mu
ABSTRACT
The endoplasmic reticulum membrane complex (EMC) plays a critical role in the biogenesis of tail-anchored proteins and a subset of multi-pass membrane proteins in the endoplasmic reticulum (ER). However, because of nearly exclusive expression of neurotransmitter-gated ion channels in the central nervous system (CNS), the role of the EMC in their biogenesis is not well understood. In this study, we demonstrated that the EMC positively regulates the surface trafficking and thus function of endogenous γ-aminobutyric acid type A (GABAA) receptors, the primary inhibitory ion channels in the mammalian brain. Moreover, among ten EMC subunits, EMC3 and EMC6 have the most prominent effect, and overexpression of EMC3 or EMC6 is sufficient to restore the function of epilepsy-associated GABAA receptor variants. In addition, EMC3 and EMC6 demonstrate endogenous interactions with major neuroreceptors, which depends on their transmembrane domains, suggesting a general role of the EMC in the biogenesis of neuroreceptors.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-GABA A Receptor β2/3 Antibody, clone 62-3G1, Upstate®, from mouse
Sigma-Aldrich
GT1-7 Mouse Hypothalamic GnRH Neuronal Cell Line, GT1-7 cells can be used as an in vitro model of GnRH-secreting neurons of the hypothalamus.
Sigma-Aldrich
Anti-GABA A Receptor α1 chain Antibody, NT, clone BD24, clone BD24, Chemicon®, from mouse
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture