MilliporeSigma
  • Reciprocal regulation of chaperone-mediated autophagy and the circadian clock.
Reciprocal regulation of chaperone-mediated autophagy and the circadian clock.
Nature cell biology (2021-12-09)
Yves R Juste, Susmita Kaushik, Mathieu Bourdenx, Ranee Aflakpui, Sanmay Bandyopadhyay, Fernando Garcia, Antonio Diaz, Kristen Lindenau, Vincent Tu, Gregory J Krause, Maryam Jafari, Rajat Singh, Javier Muñoz, Fernando Macian, Ana Maria Cuervo
ABSTRACT
Circadian rhythms align physiological functions with the light-dark cycle through oscillatory changes in the abundance of proteins in the clock transcriptional programme. Timely removal of these proteins by different proteolytic systems is essential to circadian strength and adaptability. Here we show a functional interplay between the circadian clock and chaperone-mediated autophagy (CMA), whereby CMA contributes to the rhythmic removal of clock machinery proteins (selective chronophagy) and to the circadian remodelling of a subset of the cellular proteome. Disruption of this autophagic pathway in vivo leads to temporal shifts and amplitude changes of the clock-dependent transcriptional waves and fragmented circadian patterns, resembling those in sleep disorders and ageing. Conversely, loss of the circadian clock abolishes the rhythmicity of CMA, leading to pronounced changes in the CMA-dependent cellular proteome. Disruption of this circadian clock/CMA axis may be responsible for both pathways malfunctioning in ageing and for the subsequently pronounced proteostasis defect.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Leupeptin, BioUltra, microbial, ≥95% (HPLC)