MilliporeSigma
  • Follistatin promotes LIN28B-mediated supporting cell reprogramming and hair cell regeneration in the murine cochlea.
Follistatin promotes LIN28B-mediated supporting cell reprogramming and hair cell regeneration in the murine cochlea.
Science advances (2022-02-12)
Xiao-Jun Li, Charles Morgan, Loyal A Goff, Angelika Doetzlhofer
ABSTRACT
Hair cell (HC) loss within the inner ear cochlea is a leading cause for deafness in humans. Before the onset of hearing, immature supporting cells (SCs) in neonatal mice have some limited capacity for HC regeneration. Here, we show that in organoid culture, transient activation of the progenitor-specific RNA binding protein LIN28B and Activin antagonist follistatin (FST) enhances regenerative competence of maturing/mature cochlear SCs by reprogramming them into progenitor-like cells. Transcriptome profiling and mechanistic studies reveal that LIN28B drives SC reprogramming, while FST is required to counterbalance hyperactivation of transforming growth factor-β-type signaling by LIN28B. Last, we show that LIN28B and FST coactivation enhances spontaneous cochlear HC regeneration in neonatal mice and that LIN28B may be part of an endogenous repair mechanism that primes SCs for HC regeneration. These findings indicate that SC dedifferentiation is critical for HC regeneration and identify LIN28B and FST as main regulators.
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Valproic acid sodium salt, 98%
Sigma-Aldrich
TGF-β RI Kinase Inhibitor II, TGF-β RI Kinase Inhibitor II, CAS 446859-33-2, is a cell-permeable, potent, reversible, ATP-competitive inhibitor of TGF-β R1 kinase (IC₅₀ = 23 nM and 4 nM for ALK5 binding & auto-phosphorylation).
Sigma-Aldrich
CHIR99021, ≥98% (HPLC)
Sigma-Aldrich
LY-411575, ≥97% (HPLC)
Sigma-Aldrich
Diphtheria Toxin from Corynebacterium diphtheriae, lyophilized powder, Product is in unnicked form
Sigma-Aldrich
EGF from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture