MilliporeSigma
  • Autophagy in stromal fibroblasts promotes tumor desmoplasia and mammary tumorigenesis.
Autophagy in stromal fibroblasts promotes tumor desmoplasia and mammary tumorigenesis.
Genes & development (2021-06-26)
Jenny A Rudnick, Teresa Monkkonen, Florie A Mar, James M Barnes, Hanna Starobinets, Juliet Goldsmith, Srirupa Roy, Sofía Bustamante Eguiguren, Valerie M Weaver, Jayanta Debnath
ABSTRACT
Autophagy inhibitors are currently being evaluated in clinical trials for the treatment of diverse cancers, largely due to their ability to impede tumor cell survival and metabolic adaptation. More recently, there is growing interest in whether and how modulating autophagy in the host stroma influences tumorigenesis. Fibroblasts play prominent roles in cancer initiation and progression, including depositing type 1 collagen and other extracellular matrix (ECM) components, thereby stiffening the surrounding tissue to enhance tumor cell proliferation and survival, as well as secreting cytokines that modulate angiogenesis and the immune microenvironment. This constellation of phenotypes, pathologically termed desmoplasia, heralds poor prognosis and reduces patient survival. Using mouse mammary cancer models and syngeneic transplantation assays, we demonstrate that genetic ablation of stromal fibroblast autophagy significantly impedes fundamental elements of the stromal desmoplastic response, including collagen and proinflammatory cytokine secretion, extracellular matrix stiffening, and neoangiogenesis. As a result, autophagy in stromal fibroblasts is required for mammary tumor growth in vivo, even when the cancer cells themselves remain autophagy-competent . We propose the efficacy of autophagy inhibition is shaped by this ability of host stromal fibroblast autophagy to support tumor desmoplasia.
Sigma-Aldrich
Anti-LC3 Antibody, serum, from rabbit
Sigma-Aldrich
Anti-CD45 Antibody, clone IBL-5/25, clone IBL-5/25, from rat
Sigma-Aldrich
Anti-FAP-1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®