MilliporeSigma
  • YAP, CTGF and Cyr61 are overexpressed in tamoxifen-resistant breast cancer and induce transcriptional repression of ERα.
YAP, CTGF and Cyr61 are overexpressed in tamoxifen-resistant breast cancer and induce transcriptional repression of ERα.
Journal of cell science (2021-06-08)
Hyungjoo Kim, Seogho Son, Yunhyo Ko, Jeong Eon Lee, Sangmin Kim, Incheol Shin
ABSTRACT
About 70% of breast cancers overexpress estrogen receptor α (ERα, encoded by ESR1). Tamoxifen, a competitive inhibitor of estrogen that binds to ER, has been widely used as a treatment for ER-positive breast cancer. However, 20-30% of breast cancer is resistant to tamoxifen treatment. The mechanisms underlying tamoxifen resistance remain elusive. We found that Yes-associated protein (YAP; also known as YAP1), connective tissue growth factor (CTGF; also known as CCN2) and cysteine-rich angiogenic inducer 61 (Cyr61; also known as CCN1) are overexpressed, while ERα is downregulated in tamoxifen-resistant breast cancer. Inhibition of YAP, CTGF and Cyr61 restored ERα expression and increased sensitivity to tamoxifen. Overexpression of YAP, CTGF, and Cyr61 led to downregulation of ERα and conferred resistance to tamoxifen in ER-positive breast cancer cells. Mechanistically, CTGF and Cyr61 downregulated ERα expression at the transcriptional level by directly binding to the regulatory regions of the ERα-encoding gene, leading to increased tamoxifen resistance. Also, CTGF induced Glut3 (also known as SLC2A3) expression, leading to increased glycolysis, which enhanced cell proliferation and migration in tamoxifen-resistant cells. Together, these results demonstrate a novel role of YAP, CTGF and Cyr61 in tamoxifen resistance and provide a molecular basis for their function in tamoxifen-resistant breast cancer.
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC₅₀ = 72 nM) and MEK2 (IC₅₀ = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.
Sigma-Aldrich
Verteporfin, ≥94% (HPLC)
Sigma-Aldrich
Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2, Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2, CAS 612847-09-3, is a cell-permeable, reversible inhibitor of Akt1/Akt2 (IC₅₀ = 58 nM, 210 nM, & 2.12 µM for Akt1, Akt2, and Akt3, respectively).
Sigma-Aldrich
Ribonuclease A from bovine pancreas, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
PP2, PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC₅₀ = 4, 5, 5, &100 nM for p56lck, p59fynT, Hck, & Src, respectively).
Sigma-Aldrich
Celastrol, ≥98% (HPLC), solid