MilliporeSigma
  • N-Acyl and N-sulfonyloxazolidine-2,4-diones are pseudo-irreversible inhibitors of serine proteases.
N-Acyl and N-sulfonyloxazolidine-2,4-diones are pseudo-irreversible inhibitors of serine proteases.
Bioorganic & medicinal chemistry letters (2012-05-19)
Ana Bela Santana, Susana D Lucas, Lídia M Gonçalves, Henrique F Correia, Teresa A F Cardote, Rita C Guedes, Jim Iley, Rui Moreira
ABSTRACT
The synthesis, inhibitory activity and mode of action of oxazolidine-2,4-diones against porcine pancreatic elastase, here used as a model for human neutrophil elastase, are reported. The nature of N-substitution at the oxazolidine-2,4-dione scaffold has large effect on the inhibitory potency against elastase. N-Acyl and N-sulfonyloxazolidine-2,4-diones emerged as potent pseudo-irreversible inhibitors, displaying high second-order rate constants for PPE inactivation. The title compounds were also shown to be potent inhibitors of human neutrophil elastase (HNE) and proteinase-3, and weak inhibitors of human cathepsin G. The results herein presented show that the oxazolidine-2,4-diones represent a new promising class of serine protease inhibitors.
Sigma-Aldrich
Cathepsin L Active human, recombinant, expressed in FreeStyle 293-F cells, ≥90% (SDS-PAGE)
Sigma-Aldrich
Cathepsin G from human leukocytes, lyophilized powder, ≥60 units/mg protein (Bradford)