MilliporeSigma
  • Valproate reverses mania-like behaviors in mice via preferential targeting of HDAC2.
Valproate reverses mania-like behaviors in mice via preferential targeting of HDAC2.
Molecular psychiatry (2020-11-26)
Ryan W Logan, Angela R Ozburn, Rachel N Arey, Kyle D Ketchesin, Alicia Winquist, Andrew Crain, Brian T D Tobe, Darius Becker-Krail, Matthew B Jarpe, Xiangning Xue, Wei Zong, Zhiguang Huo, Puja K Parekh, Xiyu Zhu, Ethan Fitzgerald, Hui Zhang, Jeffrey Oliver-Smith, Lauren M DePoy, Mariah A Hildebrand, Evan Y Snyder, George C Tseng, Colleen A McClung
ABSTRACT
Valproate (VPA) has been used in the treatment of bipolar disorder since the 1990s. However, the therapeutic targets of VPA have remained elusive. Here we employ a preclinical model to identify the therapeutic targets of VPA. We find compounds that inhibit histone deacetylase proteins (HDACs) are effective in normalizing manic-like behavior, and that class I HDACs (e.g., HDAC1 and HDAC2) are most important in this response. Using an RNAi approach, we find that HDAC2, but not HDAC1, inhibition in the ventral tegmental area (VTA) is sufficient to normalize behavior. Furthermore, HDAC2 overexpression in the VTA prevents the actions of VPA. We used RNA sequencing in both mice and human induced pluripotent stem cells (iPSCs) derived from bipolar patients to further identify important molecular targets. Together, these studies identify HDAC2 and downstream targets for the development of novel therapeutics for bipolar mania.
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, serum, Upstate®
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Anti-Histone H4 Antibody, pan, rabbit monoclonal, culture supernatant, clone 62-141-13, Upstate®