MilliporeSigma
  • Prolyl endopeptidase-like is a (thio)esterase involved in mitochondrial respiratory chain function.
Prolyl endopeptidase-like is a (thio)esterase involved in mitochondrial respiratory chain function.
iScience (2021-12-11)
Karen Rosier, Molly T McDevitt, Joél Smet, Brendan J Floyd, Maxime Verschoore, Maria J Marcaida, Craig A Bingman, Irma Lemmens, Matteo Dal Peraro, Jan Tavernier, Benjamin F Cravatt, Natalia V Gounko, Katlijn Vints, Yenthe Monnens, Kritika Bhalla, Laetitia Aerts, Edrees H Rashan, Arnaud V Vanlander, Rudy Van Coster, Luc Régal, David J Pagliarini, John W M Creemers
ABSTRACT
Deficiency of the serine hydrolase prolyl endopeptidase-like (PREPL) causes a recessive metabolic disorder characterized by neonatal hypotonia, feeding difficulties, and growth hormone deficiency. The pathophysiology of PREPL deficiency and the physiological substrates of PREPL remain largely unknown. In this study, we connect PREPL with mitochondrial gene expression and oxidative phosphorylation by analyzing its protein interactors. We demonstrate that the long PREPLL isoform localizes to mitochondria, whereas PREPLS remains cytosolic. Prepl KO mice showed reduced mitochondrial complex activities and disrupted mitochondrial gene expression. Furthermore, mitochondrial ultrastructure was abnormal in a PREPL-deficient patient and Prepl KO mice. In addition, we reveal that PREPL has (thio)esterase activity and inhibition of PREPL by Palmostatin M suggests a depalmitoylating function. We subsequently determined the crystal structure of PREPL, thereby providing insight into the mechanism of action. Taken together, PREPL is a (thio)esterase rather than a peptidase and PREPLL is involved in mitochondrial homeostasis.
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% based on Mol. Wt. 12,384 basis
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
APT1 Inhibitor, palmostatin B, The APT1 Inhibitor, palmostatin B controls the biological activity of APT1.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
4-Nitrophenyl dodecanoate, ≥98.0% (GC)