MilliporeSigma
  • Synthesis of Either C2- or C4'-Alkylated Derivatives of Honokiol and Their Biological Evaluation for Anti-inflammatory Activity.
Synthesis of Either C2- or C4'-Alkylated Derivatives of Honokiol and Their Biological Evaluation for Anti-inflammatory Activity.
Chemical & pharmaceutical bulletin (2019-07-02)
San-Ha Lee, Xiang Fei, Chaelin Lee, Hien Thi Thu Do, Inmoo Rhee, Seung-Yong Seo
ABSTRACT
Honokiol, a biphenolic neolignan isolated from Magnolia officinalis, was reported to have a promising anti-inflammatory activity for the treatment of various diseases. There are many efforts on the synthesis and structure-activity relationship of honokiol derivatives. However, regioselective O-alkylation of honokiol remains a challenge and serves as a tool to provide not only some derivatives but also chemical probes for target identification and mode of action. In this study, we examined the reaction condition for regioselective O-alkylation, in which C2 and C4'-alkylated analogs of honokiol were synthesized and evaluated for inhibitory activity on nitric oxide production and cyclooxygenase-2 expression. Furthermore, we successfully synthesized a potential photoaffinity probe consisting of biotin and benzophenone based on a C4'-alkylated derivative.
Product Description
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Millipore
NP-40 Alternative