MilliporeSigma
  • Activators of alpha synuclein expression identified by reporter cell line-based high throughput drug screen.
Activators of alpha synuclein expression identified by reporter cell line-based high throughput drug screen.
Scientific reports (2021-10-08)
Fabian Stahl, Philip Denner, Dominik Piston, Bernd O Evert, Laura de Boni, Ina Schmitt, Peter Breuer, Ullrich Wüllner
ABSTRACT
Multiplications, mutations and dysregulation of the alpha synuclein gene (SNCA) are associated with the demise of dopaminergic neurons and are considered to play important roles in the pathogenesis of familial and sporadic forms of Parkinson's disease. Regulation of SNCA expression might thus be an appropriate target for treatment. We aimed to identify specific modulators of SNCA transcription, generated CRISPR/Cas9 modified SNCA-GFP-luciferase (LUC) genomic fusion- and control cell lines and screened a library of 1649 bioactive compounds, including the FDA approved drugs. We found no inhibitors but three selective activators which increased SNCA mRNA and protein levels.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-α-Synuclein Antibody, clone 2F12, clone 2F12, from mouse
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, 1 mg/mL, Upstate®