MilliporeSigma
  • Nuclear envelope-localized torsinA-LAP1 complex regulates hepatic VLDL secretion and steatosis.
Nuclear envelope-localized torsinA-LAP1 complex regulates hepatic VLDL secretion and steatosis.
The Journal of clinical investigation (2019-08-14)
Ji-Yeon Shin, Antonio Hernandez-Ono, Tatyana Fedotova, Cecilia Östlund, Michael J Lee, Sarah B Gibeley, Chun-Chi Liang, William T Dauer, Henry N Ginsberg, Howard J Worman
ABSTRACT
Deciphering novel pathways regulating liver lipid content has profound implications for understanding the pathophysiology of nonalcoholic fatty liver disease and nonalcoholic steatohepatitis. Recent evidence suggests that the nuclear envelope is a site of regulation of lipid metabolism but there is limited appreciation of the responsible mechanisms and molecular components within this organelle. We showed that conditional hepatocyte deletion of the inner nuclear membrane protein lamina-associated polypeptide 1 (LAP1) caused defective VLDL secretion and steatosis, including intranuclear lipid accumulation. LAP1 binds to and activates torsinA, an AAA+ ATPase that resides in the perinuclear space and continuous main ER. Deletion of torsinA from mouse hepatocytes caused even greater reductions in VLDL secretion and profound steatosis. Both of these mutant mouse lines developed hepatic steatosis and subsequent steatohepatitis on a regular chow diet in the absence of whole-body insulin resistance or obesity. Our results establish an essential role for the nuclear envelope-localized torsinA-LAP1 complex in hepatic VLDL secretion and suggest that the torsinA pathway participates in the pathophysiology of nonalcoholic fatty liver disease.
Sigma-Aldrich
BCG Albumin Assay Kit, sufficient for 250 colorimetric tests
Sigma-Aldrich
Anti-Apolipoprotein B Goat pAb, liquid, Calbiochem®