MilliporeSigma
  • Commensal fungi and their cell-wall β-glucans direct differential responses in human intestinal epithelial cells.
Commensal fungi and their cell-wall β-glucans direct differential responses in human intestinal epithelial cells.
European journal of immunology (2021-02-23)
Sarit Cohen-Kedar, Danielle Keizer, Suzana Schwartz, Keren M Rabinowitz, Kawsar Kaboub, Efrat Shaham Barda, Eran Sadot, Meirav Wolff-Bar, Tali Shaltiel, Iris Dotan
ABSTRACT
Intestinal epithelial cells (IECs) are the first to encounter luminal antigens and play an active role in intestinal immune responses. We previously reported that β-glucans, major fungal cell-wall glycans, induced chemokine secretion by IEC lines in a Dectin-1- and Syk-dependent manner. Here, we show that in contrast to β-glucans, stimulation of IEC lines with Candida albicans and Saccharomyces cerevisiae did not induce secretion of any of the proinflammatory cytokines IL-8, CCL2, CXCL1, and GM-CSF. Commensal fungi and β-glucans activated Syk and ERK in IEC lines. However, only β-glucans activated p38, JNK, and the transcription factors NF-κB p65 and c-JUN, which were necessary for cytokine secretion. Furthermore, costimulation of IEC lines with β-glucans and C. albicans yielded decreased cytokine secretion compared to stimulation with β-glucans alone. Finally, ex vivo stimulation of human colonic mucosal explants with zymosan and C. albicans, leads to epithelial Syk and ERK phosphorylation, implying recognition of fungi and similar initial signaling pathways as in IEC lines. Lack of cytokine secretion in response to commensal fungi may reflect IECs' response to fungal glycans, other than β-glucans, that contribute to mucosal tolerance. Skewed epithelial response to commensal fungi may impair homeostasis and contribute to intestinal inflammation.
Sigma-Aldrich
Syk Inhibitor, The Spleen Tyrosine Kinase Inhibitor, also referenced under CAS 622387-85-3, controls the biological activity of Spleen Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC₅₀ = 72 nM) and MEK2 (IC₅₀ = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.
Sigma-Aldrich
Protease Inhibitor Cocktail Set I, A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.
Millipore
Phosphatase Inhibitor Cocktail Set II, A cocktail of five phosphatase inhibitors for the inhibition of acid and alkaline phosphatases as well as protein tyrosine phosphatases (PTPs). Suitable for use with cell lysates and tissue extracts.
Sigma-Aldrich
Bay 11-7082, ≥98% (HPLC), powder
Sigma-Aldrich
Curdlan from Alcaligenes faecalis