MilliporeSigma
  • Cell-type-specific profiling of human cellular models of fragile X syndrome reveal PI3K-dependent defects in translation and neurogenesis.
Cell-type-specific profiling of human cellular models of fragile X syndrome reveal PI3K-dependent defects in translation and neurogenesis.
Cell reports (2021-04-15)
Nisha Raj, Zachary T McEachin, William Harousseau, Ying Zhou, Feiran Zhang, Megan E Merritt-Garza, J Matthew Taliaferro, Magdalena Kalinowska, Samuele G Marro, Chadwick M Hales, Elizabeth Berry-Kravis, Marisol W Wolf-Ochoa, Veronica Martinez-Cerdeño, Marius Wernig, Lu Chen, Eric Klann, Stephen T Warren, Peng Jin, Zhexing Wen, Gary J Bassell
ABSTRACT
Transcriptional silencing of the FMR1 gene in fragile X syndrome (FXS) leads to the loss of the RNA-binding protein FMRP. In addition to regulating mRNA translation and protein synthesis, emerging evidence suggests that FMRP acts to coordinate proliferation and differentiation during early neural development. However, whether loss of FMRP-mediated translational control is related to impaired cell fate specification in the developing human brain remains unknown. Here, we use human patient induced pluripotent stem cell (iPSC)-derived neural progenitor cells and organoids to model neurogenesis in FXS. We developed a high-throughput, in vitro assay that allows for the simultaneous quantification of protein synthesis and proliferation within defined neural subpopulations. We demonstrate that abnormal protein synthesis in FXS is coupled to altered cellular decisions to favor proliferative over neurogenic cell fates during early development. Furthermore, pharmacologic inhibition of elevated phosphoinositide 3-kinase (PI3K) signaling corrects both excess protein synthesis and cell proliferation in a subset of patient neural cells.
Sigma-Aldrich
Anti-TRA-1-81 Antibody, clone TRA-1-81, clone TRA-1-81, Chemicon®, from mouse
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse
Sigma-Aldrich
Anti-PI3 Kinase Antibody, p110β, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Puromycin, clone 12D10, Alexa Fluor 488 Conjugate Antibody, clone 12D10, 0.5 mg/mL, from mouse
Roche
Cell Proliferation ELISA, BrdU (colorimetric), sufficient for ≤1,000 tests
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse