Merck
  • Short Access to Belt Compounds with Spatially Close C=C Bonds and Their Transannular Reactions.
Short Access to Belt Compounds with Spatially Close C=C Bonds and Their Transannular Reactions.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-09-17)
Pelayo Camps, Tània Gómez, Ane Otermin, Mercè Font-Bardia, Carolina Estarellas, Francisco Javier Luque
ABSTRACT
Two domino Diels-Alder adducts were obtained from 3,7-bis(cyclopenta-2,4-dien-1-ylidene)-cis-bicyclo[3.3.0]octane and dimethyl acetylenedicarboxylate or N-methylmaleimide under microwave irradiation. From the first adduct, a C20H24 diene with C2v symmetry was obtained by Zn/AcOH reduction, hydrolysis, oxidative decarboxylation, and selective hydrogenation. Photochemical [2+2] cycloaddition of this diene gave a thermally unstable cyclobutane derivative, which reverts to the diene. However, both the diene and the cyclobutane derivatives could be identified by X-ray diffraction analysis upon irradiation of the diene crystal. New six-membered rings are formed upon the transannular addition of bromine or iodine to the diene. The N-type selectivity of the addition was examined by theoretical calculations, which revealed the distinct susceptibility of the doubly bonded carbon atoms to the bromine attack.
Sigma-Aldrich
Anti-KDR/Flk-1/VEGFR2 Antibody, clone CH-11, clone CH-11, Upstate®, from mouse