MilliporeSigma
  • STAT3-dependent regulation of the electrogenic Na+/ HCO3 - cotransporter 1 (NBCe1) functional expression in cortical astrocytes.
STAT3-dependent regulation of the electrogenic Na+/ HCO3 - cotransporter 1 (NBCe1) functional expression in cortical astrocytes.
Journal of cellular physiology (2020-08-08)
Marina Giannaki, Magdalena Schrödl-Häußel, Shokoufeh Khakipoor, Matthias Kirsch, Eleni Roussa
ABSTRACT
The electrogenic Na+ /HCO3 - cotransporter (NBCe1) in astrocytes is crucial in regulation of acid-base homeostasis in the brain. Since many pathophysiological conditions in the brain have been associated with pH shifts we exposed primary mouse cortical and hippocampal astrocytes to prolonged low or high extracellular pH (pHo ) at constant extracellular bicarbonate concentration and investigated activation of astrocytes and regulation of NBCe1 by immunoblotting, biotinylation of surface proteins, and intracellular H+ recordings. High pHo at constant extracellular bicarbonate caused upregulation of NBCe1 protein, surface expression and activity via upregulation of the astrocytic activation markers signal transducer and activator of transcription 3 (STAT3) signaling and glial fibrillary acidic protein expression. High pHo -induced increased NBCe1 protein expression was prevented in astrocytes from Stat3flox/flox ::GfapCre/+ mice. In vitro, basal and high pHo -induced increased NBCe1 functional expression was impaired following inhibition of STAT3 phosphorylation. These results provide a novel regulation mode of NBCe1 protein and activity, highlight the importance of astrocyte reactivity on regulation of NBCe1 and implicate roles for NBCe1 in altering/modulating extracellular pH during development as well as of the microenvironment at sites of brain injuries and other pathophysiological conditions.
Sigma-Aldrich
PP2, PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC₅₀ = 4, 5, 5, &100 nM for p56lck, p59fynT, Hck, & Src, respectively).
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder