MilliporeSigma
  • BAX-dependent mitochondrial pathway mediates the crosstalk between ferroptosis and apoptosis.
BAX-dependent mitochondrial pathway mediates the crosstalk between ferroptosis and apoptosis.
Apoptosis : an international journal on programmed cell death (2020-08-02)
Young-Sun Lee, Kalishwaralal Kalimuthu, Yong Seok Park, Xu Luo, M Haroon A Choudry, David L Bartlett, Yong J Lee
ABSTRACT
Ferroptosis is considered a distinctive form of cell death compared to other types of death such as apoptosis. It is known to result from iron-dependent accumulation of lipid peroxides rather than caspase activation. However, we reported recently that ferroptosis interplays with apoptosis. In this study, we investigated a possible mechanism of this interplay between ferroptosis and apoptosis. Results from our studies reveal that combined treatment of the ferroptotic agent erastin and the apoptotic agent TRAIL effectively disrupted mitochondrial membrane potential (ΔΨm) and subsequently promoted caspase activation. The alterations of mitochondrial membrane potential are probably due to an increase in oligomerization of BAX and its accumulation at the mitochondria during treatment with erastin and TRAIL. Interestingly, the combined treatment-promoted apoptosis was effectively inhibited in BAX-deficient HCT116 cells, but not BAK-deficient cells. These results indicate that the BAX-associated mitochondria-dependent pathway plays a pivotal role in erastin-enhanced TRAIL-induced apoptosis.
Product Description
Supelco
N-(Propionyloxy)succinimide, for LC-MS derivatization, LiChropur, ≥95% (HPLC)