MilliporeSigma
  • Identification of candidate host serum and saliva biomarkers for a better diagnosis of active and latent tuberculosis infection.
Identification of candidate host serum and saliva biomarkers for a better diagnosis of active and latent tuberculosis infection.
PloS one (2020-07-21)
Olivia Estévez, Luis Anibarro, Elina Garet, Ángeles Pallares, Alberto Pena, Carlos Villaverde, Víctor Del Campo, África González-Fernández
ABSTRACT
In our work, we aim to identify new candidate host biomarkers to discriminate between active TB patients (n = 28), latent infection (LTBI; n = 27) and uninfected (NoTBI; n = 42) individuals. For that, active TB patients and their contacts were recruited that donated serum and saliva samples. A multiplex assay was performed to study the concentration of different cytokines, chemokines and growth factors. Proteins with significant differences between groups were selected and logistic regression and the area under the ROC curve (AUC) was used to assess the diagnostic accuracy. The best marker combinations that discriminate active TB from NoTBI contacts were [IP-10 + IL-7] in serum and [Fractalkine + IP-10 + IL-1α + VEGF] in saliva. Best discrimination between active TB and LTBI was achieved using [IP-10 + BCA-1] in serum (AUC = 0.83) and IP-10 in saliva (p = 0.0007; AUC = 0.78). The levels of TNFα (p = 0.003; AUC = 0.73) in serum and the combination of [Fractalkine+IL-12p40] (AUC = 0.83) in saliva, were able to differentiate between NoTBI and LTBI contacts. In conclusion, different individual and combined protein markers could help to discriminate between active TB and both uninfected and latently-infected contacts. The most promising ones include [IP-10 + IL-7], [IP-10 + BCA-1] and TNFα in serum and [Fractalkine + IP-10 + IL-1α + VEGF], IP-10 and [Fractalkine+IL-12p40] in saliva.
Product Description
Millipore
MILLIPLEX® Human Cytokine/Chemokine Magnetic Bead Panel II - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.
Millipore
MILLIPLEX® Human TH17 Magnetic Bead Panel - Immunology Multiplex Assay, Simultaneously analyze multiple Th17 cytokine and chemokine biomarkers with the Th17 Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.