MilliporeSigma
  • Characterization of the activity of the PI3K/mTOR inhibitor XL765 (SAR245409) in tumor models with diverse genetic alterations affecting the PI3K pathway.
Characterization of the activity of the PI3K/mTOR inhibitor XL765 (SAR245409) in tumor models with diverse genetic alterations affecting the PI3K pathway.
Molecular cancer therapeutics (2014-03-19)
Peiwen Yu, A Douglas Laird, Xiangnan Du, Jianming Wu, Kwang-Ai Won, Kyoko Yamaguchi, Pin Pin Hsu, Fawn Qian, Christopher T Jaeger, Wentao Zhang, Chris A Buhr, Paula Shen, Wendy Abulafia, Jason Chen, Jenny Young, Arthur Plonowski, F Michael Yakes, Felix Chu, Michelle Lee, Frauke Bentzien, Sanh Tan Lam, Stephanie Dale, David J Matthews, Peter Lamb, Paul Foster
ABSTRACT
Activation of the PI3K (phosphoinositide 3-kinase) pathway is a frequent occurrence in human tumors and is thought to promote growth, survival, and resistance to diverse therapies. Here, we report pharmacologic characterization of the pyridopyrimidinone derivative XL765 (SAR245409), a potent and highly selective pan inhibitor of class I PI3Ks (α, β, γ, and δ) with activity against mTOR. Broad kinase selectivity profiling of >130 protein kinases revealed that XL765 is highly selective for class I PI3Ks and mTOR over other kinases. In cellular assays, XL765 inhibits the formation of PIP(3) in the membrane, and inhibits phosphorylation of AKT, p70S6K, and S6 phosphorylation in multiple tumor cell lines with different genetic alterations affecting the PI3K pathway. In a panel of tumor cell lines, XL765 inhibits proliferation with a wide range of potencies, with evidence of an impact of genotype on sensitivity. In mouse xenograft models, oral administration of XL765 results in dose-dependent inhibition of phosphorylation of AKT, p70S6K, and S6 with a duration of action of approximately 24 hours. Repeat dose administration of XL765 results in significant tumor growth inhibition in multiple human xenograft models in nude mice that is associated with antiproliferative, antiangiogenic, and proapoptotic effects.
Product Description
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-Cyclin D1 (Ab-3) Mouse mAb (DCS-6), liquid, clone DCS-6, Calbiochem®
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)