MilliporeSigma
  • Plant-derived exosomal microRNAs inhibit lung inflammation induced by exosomes SARS-CoV-2 Nsp12.
Plant-derived exosomal microRNAs inhibit lung inflammation induced by exosomes SARS-CoV-2 Nsp12.
Molecular therapy : the journal of the American Society of Gene Therapy (2021-05-14)
Yun Teng, Fangyi Xu, Xiangcheng Zhang, Jingyao Mu, Mohammed Sayed, Xin Hu, Chao Lei, Mukesh Sriwastva, Anil Kumar, Kumaran Sundaram, Lifeng Zhang, Juw Won Park, Shao-Yu Chen, Shuangqin Zhang, Jun Yan, Michael L Merchant, Xiang Zhang, Craig J McClain, Jennifer K Wolfe, Robert S Adcock, Donghoon Chung, Kenneth E Palmer, Huang-Ge Zhang
ABSTRACT
Lung inflammation is a hallmark of coronavirus disease 2019 (COVID-19). In this study, we show that mice develop inflamed lung tissue after being administered exosomes released from the lung epithelial cells exposed to severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) Nsp12 and Nsp13 (exosomesNsp12Nsp13). Mechanistically, we show that exosomesNsp12Nsp13 are taken up by lung macrophages, leading to activation of nuclear factor κB (NF-κB) and the subsequent induction of an array of inflammatory cytokines. Induction of tumor necrosis factor (TNF)-α, interleukin (IL)-6, and IL-1β from exosomesNsp12Nsp13-activated lung macrophages contributes to inducing apoptosis in lung epithelial cells. Induction of exosomesNsp12Nsp13-mediated lung inflammation was abolished with ginger exosome-like nanoparticle (GELN) microRNA (miRNA aly-miR396a-5p. The role of GELNs in inhibition of the SARS-CoV-2-induced cytopathic effect (CPE) was further demonstrated via GELN aly-miR396a-5p- and rlcv-miR-rL1-28-3p-mediated inhibition of expression of Nsp12 and spike genes, respectively. Taken together, our results reveal exosomesNsp12Nsp13 as potentially important contributors to the development of lung inflammation, and GELNs are a potential therapeutic agent to treat COVID-19.
Sigma-Aldrich
Anti-FLAG Tag antibody produced in rabbit, affinity isolated antibody