MilliporeSigma
  • Design, Optimization, and Study of Small Molecules That Target Tau Pre-mRNA and Affect Splicing.
Design, Optimization, and Study of Small Molecules That Target Tau Pre-mRNA and Affect Splicing.
Journal of the American Chemical Society (2020-05-05)
Jonathan L Chen, Peiyuan Zhang, Masahito Abe, Haruo Aikawa, Liying Zhang, Alexander J Frank, Timothy Zembryski, Christopher Hubbs, HaJeung Park, Jane Withka, Claire Steppan, Lucy Rogers, Shawn Cabral, Martin Pettersson, Travis T Wager, Matthew A Fountain, Gavin Rumbaugh, Jessica L Childs-Disney, Matthew D Disney
ABSTRACT
Approximately 95% of human genes are alternatively spliced, and aberrant splicing events can cause disease. One pre-mRNA that is alternatively spliced and linked to neurodegenerative diseases is tau (microtubule-associated protein tau), which can cause frontotemporal dementia and parkinsonism linked to chromosome 17 (FTDP-17) and can contribute to Alzheimer's disease. Here, we describe the design of structure-specific lead small molecules that directly target tau pre-mRNA from sequence. This was followed by hit expansion and analogue synthesis to further improve upon these initial lead molecules. The emergent compounds were assessed for functional activity in a battery of assays, including binding assays and an assay that mimics molecular recognition of tau pre-mRNA by a U1 small nuclear ribonucleoprotein (snRNP) splicing factor. Compounds that emerged from these studies had enhanced potency and selectivity for the target RNA relative to the initial hits, while also having significantly improved drug-like properties. The compounds are shown to directly target tau pre-mRNA in cells, via chemical cross-linking and isolation by pull-down target profiling, and to rescue disease-relevant splicing of tau pre-mRNA in a variety of cellular systems, including primary neurons. More broadly, this study shows that lead, structure-specific compounds can be designed from sequence and then further optimized for their physicochemical properties while at the same time enhancing their activity.
Sigma-Aldrich
Anti-Tau (3-repeat isoform RD3) Antibody, clone 8E6/C11, culture supernatant, clone 8E6/C11, Upstate®
Sigma-Aldrich
Anti-Tau (4-repeat isoform RD4) Antibody, clone 1E1/A6, culture supernatant, clone 1E1/A6, Upstate®
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Protease Inhibitor Cocktail Set V, EDTA-Free, A cocktail of four protease inhibitors for the inhibition of serine, cysteine, but not metalloproteases.
Sigma-Aldrich
Poly-D-lysine hydrobromide, average mol wt 30,000-70,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture