MilliporeSigma
  • Development of New Modified Simple Polymerase Chain Reaction and Real-time Polymerase Chain Reaction for the Identification of Iranian Brucella abortus Strains.
Development of New Modified Simple Polymerase Chain Reaction and Real-time Polymerase Chain Reaction for the Identification of Iranian Brucella abortus Strains.
Archives of Razi Institute (2019-10-09)
S Alamian, T Zahraei Salehi, K Aghaiypour Kolyani, M Esmaelizad, A Etemadi
ABSTRACT
Brucellosis is primarily a worldwide zoonotic disease caused by Brucella species. The genus Brucella contains highly infectious species that are classified as biological threat agents. In this regard, the identification of Brucella can be a time-consuming and labor-intensive process posing a real risk of laboratory-acquired infection to the laboratory staff. This study aimed to present a novel conventional and real-time polymerase chain reaction (PCR) assay for the identification of Brucella abortus strains. Regarding this, two primers (bru ab2) were designed based on the unique loci encoding autotransporter-associated beta strand repeat-containing protein (ID:YP00113760). A total of 56 Brucella strains (e.g., reference, vaccinal, and field isolates) and Yersinia enterocolitica, as a non-Brucella isolate, were evaluated in conventional and real-time PCR systems. The results of the study indicated that 0.4 ng and 400 FG of genomic DNA of B. abortus strains can be detected by conventional and real-time PCR, respectively. The primers, bru ab2, were suitable for both PCR methods. Both methods were specific for the detection of all strains of the bacterium; however, real-time PCR assay was 1000-fold more sensitive than the conventional PCR method. Therefore, this new detection system could be a suitable selective modified method for the accurate identification of all B. abortus strains.
Product Description
Roche
High Pure PCR Template Preparation Kit, pkg of 100 purifications, suitable for DNA extraction