MilliporeSigma
  • Integrin-independent tyrosine phosphorylation of p125(fak) in human platelets stimulated by collagen.
Integrin-independent tyrosine phosphorylation of p125(fak) in human platelets stimulated by collagen.
The Journal of biological chemistry (2000-12-09)
M Achison, C M Elton, P G Hargreaves, C G Knight, M J Barnes, R W Farndale
ABSTRACT
Collagen fibers or a glycoprotein VI-specific collagen-related peptide (CRP-XL) stimulated tyrosine phosphorylation of the focal adhesion kinase, p125(fak) (FAK), in human platelets. An integrin alpha(2)beta(1)-specific triple-helical peptide ligand, containing the sequence GFOGER (single-letter nomenclature, O = Hyp) was without effect. Antibodies to the alpha(2) and beta(1) integrin subunits did not inhibit platelet FAK tyrosine phosphorylation caused by either collagen fibers or CRP-XL. Tyrosine phosphorylation of FAK caused by CRP-XL or thrombin, but not that caused by collagen fibers, was partially inhibited by GR144053F, an antagonist of integrin alpha(IIb)beta(3). The intracellular Ca(2+) chelator, BAPTA, and the protein kinase C inhibitor, Ro31-8220, were each highly effective inhibitors of the FAK tyrosine phosphorylation caused by collagen or CRP-XL. These data suggest that, in human platelets, 1) occupation or clustering of the integrin alpha(2)beta(1) is neither sufficient nor necessary for activation of FAK, 2) the fibrinogen receptor alpha(IIb)beta(3) is not required for activation of FAK by collagen fibers, and 3) both intracellular Ca(2+) and protein kinase C activity are essential intermediaries of FAK activation.
Sigma-Aldrich
Ro-31-8220, A cell-permeable, reversible, competitive, and selective inhibitor of protein kinase C (PKC; IC₅₀ = 10 nM) over CaM kinase II (IC₅₀ = 17 µM) and protein kinase A (IC₅₀ = 900 nM).
Sigma-Aldrich
PANSORBIN® Cells
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Millipore
BAPTA/AM, Membrane-permeable form of BAPTA.