MilliporeSigma
  • Integrative Analysis Identifies Key Molecular Signatures Underlying Neurodevelopmental Deficits in Fragile X Syndrome.
Integrative Analysis Identifies Key Molecular Signatures Underlying Neurodevelopmental Deficits in Fragile X Syndrome.
Biological psychiatry (2020-07-13)
Kagistia Hana Utami, Niels H Skotte, Ana R Colaço, Nur Amirah Binte Mohammad Yusof, Bernice Sim, Xin Yi Yeo, Han-Gyu Bae, Marta Garcia-Miralles, Carola I Radulescu, Qiyu Chen, Georgia Chaldaiopoulou, Herty Liany, Srikanth Nama, Ulla-Kaisa A Peteri, Prabha Sampath, Maija L Castrén, Sangyong Jung, Matthias Mann, Mahmoud A Pouladi
ABSTRACT
Fragile X syndrome (FXS) is a neurodevelopmental disorder caused by epigenetic silencing of FMR1 and loss of FMRP expression. Efforts to understand the molecular underpinnings of the disease have been largely performed in rodent or nonisogenic settings. A detailed examination of the impact of FMRP loss on cellular processes and neuronal properties in the context of isogenic human neurons remains lacking. Using CRISPR (clustered regularly interspaced short palindromic repeats)/Cas9 to introduce indels in exon 3 of FMR1, we generated an isogenic human pluripotent stem cell model of FXS that shows complete loss of FMRP expression. We generated neuronal cultures and performed genome-wide transcriptome and proteome profiling followed by functional validation of key dysregulated processes. We further analyzed neurodevelopmental and neuronal properties, including neurite length and neuronal activity, using multielectrode arrays and patch clamp electrophysiology. We showed that the transcriptome and proteome profiles of isogenic FMRP-deficient neurons demonstrate perturbations in synaptic transmission, neuron differentiation, cell proliferation and ion transmembrane transporter activity pathways, and autism spectrum disorder-associated gene sets. We uncovered key deficits in FMRP-deficient cells demonstrating abnormal neural rosette formation and neural progenitor cell proliferation. We further showed that FMRP-deficient neurons exhibit a number of additional phenotypic abnormalities, including neurite outgrowth and branching deficits and impaired electrophysiological network activity. These FMRP-deficient related impairments have also been validated in additional FXS patient-derived human-induced pluripotent stem cell neural cells. Using isogenic human pluripotent stem cells as a model to investigate the pathophysiology of FXS in human neurons, we reveal key neural abnormalities arising from the loss of FMRP.
Sigma-Aldrich
Anti-Ki-67 Antibody, clone Ki-S5, clone Ki-S5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Microtubule-Associated Protein 2 (MAP2) Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-4 Antibody, clone MC-813-70, clone MC-813-70, Chemicon®, from mouse
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Actin Antibody, smooth muscle, clone ASM-1, clone ASM-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-AFP Mouse mAb (1G7), liquid, clone 1G7, Calbiochem®
Sigma-Aldrich
Anti-Fragile X Mental Retardation Protein Antibody, clone 1C3, ascites fluid, clone 1C3, Chemicon®
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®
Sigma-Aldrich
Anti-Nestin Antibody, clone 10C2, clone 10C2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Calnexin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder