MilliporeSigma
The ERBB network facilitates KRAS-driven lung tumorigenesis.
Science translational medicine (2018-06-22)
Björn Kruspig, Tiziana Monteverde, Sarah Neidler, Andreas Hock, Emma Kerr, Colin Nixon, William Clark, Ann Hedley, Sarah Laing, Seth B Coffelt, John Le Quesne, Craig Dick, Karen H Vousden, Carla P Martins, Daniel J Murphy
ABSTRACT
KRAS is the most frequently mutated driver oncogene in human adenocarcinoma of the lung. There are presently no clinically proven strategies for treatment of KRAS-driven lung cancer. Activating mutations in KRAS are thought to confer independence from upstream signaling; however, recent data suggest that this independence may not be absolute. We show that initiation and progression of KRAS-driven lung tumors require input from ERBB family receptor tyrosine kinases (RTKs): Multiple ERBB RTKs are expressed and active from the earliest stages of KRAS-driven lung tumor development, and treatment with a multi-ERBB inhibitor suppresses formation of KRASG12D-driven lung tumors. We present evidence that ERBB activity amplifies signaling through the core RAS pathway, supporting proliferation of KRAS-mutant tumor cells in culture and progression to invasive disease in vivo. Brief pharmacological inhibition of the ERBB network enhances the therapeutic benefit of MEK (mitogen-activated protein kinase kinase) inhibition in an autochthonous tumor setting. Our data suggest that lung cancer patients with KRAS-driven disease may benefit from inclusion of multi-ERBB inhibitors in rationally designed treatment strategies.
Sigma-Aldrich
Anti-phospho-erbB-2/HER-2 (Tyr1248) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-c-ErbB2/c-Neu (Ab-3) Mouse mAb (3B5), lyophilized, clone 3B5, Calbiochem®
Sigma-Aldrich
Anti-EGFR Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-erbB-3/HER-3 Antibody, clone 2F12, clone 2F12, Upstate®, from mouse