MilliporeSigma
  • ACE2 localizes to the respiratory cilia and is not increased by ACE inhibitors or ARBs.
ACE2 localizes to the respiratory cilia and is not increased by ACE inhibitors or ARBs.
Nature communications (2020-10-30)
Ivan T Lee, Tsuguhisa Nakayama, Chien-Ting Wu, Yury Goltsev, Sizun Jiang, Phillip A Gall, Chun-Kang Liao, Liang-Chun Shih, Christian M Schürch, David R McIlwain, Pauline Chu, Nicole A Borchard, David Zarabanda, Sachi S Dholakia, Angela Yang, Dayoung Kim, Han Chen, Tomoharu Kanie, Chia-Der Lin, Ming-Hsui Tsai, Katie M Phillips, Raymond Kim, Jonathan B Overdevest, Matthew A Tyler, Carol H Yan, Chih-Feng Lin, Yi-Tsen Lin, Da-Tian Bau, Gregory J Tsay, Zara M Patel, Yung-An Tsou, Alexandar Tzankov, Matthias S Matter, Chih-Jaan Tai, Te-Huei Yeh, Peter H Hwang, Garry P Nolan, Jayakar V Nayak, Peter K Jackson
ABSTRACT
The coronavirus SARS-CoV-2 is the causative agent of the ongoing severe acute respiratory disease pandemic COVID-19. Tissue and cellular tropism is one key to understanding the pathogenesis of SARS-CoV-2. We investigate the expression and subcellular localization of the SARS-CoV-2 receptor, angiotensin-converting enzyme 2 (ACE2), within the upper (nasal) and lower (pulmonary) respiratory tracts of human donors using a diverse panel of banked tissues. Here, we report our discovery that the ACE2 receptor protein robustly localizes within the motile cilia of airway epithelial cells, which likely represents the initial or early subcellular site of SARS-CoV-2 viral entry during host respiratory transmission. We further determine whether ciliary ACE2 expression in the upper airway is influenced by patient demographics, clinical characteristics, comorbidities, or medication use, and show the first mechanistic evidence that the use of angiotensin-converting enzyme inhibitors (ACEI) or angiotensin II receptor blockers (ARBs) does not increase susceptibility to SARS-CoV-2 infection through enhancing the expression of ciliary ACE2 receptor. These findings are crucial to our understanding of the transmission of SARS-CoV-2 for prevention and control of this virulent pathogen.
Sigma-Aldrich
Anti-FMRFamide Antibody, serum, Chemicon®
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Anti-ACE2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution