MilliporeSigma
  • The tomato receptor CuRe1 senses a cell wall protein to identify Cuscuta as a pathogen.
The tomato receptor CuRe1 senses a cell wall protein to identify Cuscuta as a pathogen.
Nature communications (2020-10-22)
Volker Hegenauer, Peter Slaby, Max Körner, Julien-Alexander Bruckmüller, Ronja Burggraf, Isabell Albert, Bettina Kaiser, Birgit Löffelhardt, Irina Droste-Borel, Jan Sklenar, Frank L H Menke, Boris Maček, Aashish Ranjan, Neelima Sinha, Thorsten Nürnberger, Georg Felix, Kirsten Krause, Mark Stahl, Markus Albert
ABSTRACT
Parasitic plants of the genus Cuscuta penetrate shoots of host plants with haustoria and build a connection to the host vasculature to exhaust water, solutes and carbohydrates. Such infections usually stay unrecognized by the host and lead to harmful host plant damage. Here, we show a molecular mechanism of how plants can sense parasitic Cuscuta. We isolated an 11 kDa protein of the parasite cell wall and identified it as a glycine-rich protein (GRP). This GRP, as well as its minimal peptide epitope Crip21, serve as a pathogen-associated molecular pattern and specifically bind and activate a membrane-bound immune receptor of tomato, the Cuscuta Receptor 1 (CuRe1), leading to defense responses in resistant hosts. These findings provide the initial steps to understand the resistance mechanisms against parasitic plants and further offer great potential for protecting crops by engineering resistance against parasitic plants.
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Roche
Streptavidin-AP conjugate, solution, pkg of 1 mL (1,000U)
Sigma-Aldrich
Anti-c-Myc antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution