- MicroRNA-572/hMOF/Sirt6 regulates the progression of ovarian cancer.
MicroRNA-572/hMOF/Sirt6 regulates the progression of ovarian cancer.
Human males absent on the first (hMOF) is a histone acetyltransferase (HAT) and is involved in the pathogenesis of various cancers. This article aimed to reveal the potential mechanism of the miR-572/hMOF/Sirt6 axis in ovarian cancer (OC). In this study, we found that the mRNA and protein levels of hMOF and Sirt6 were abnormally down-regulated in OC tissues and cells. Further study indicated that the overexpression of hMOF increased the level of H4 histone acetylation in the Sirt6 promoter region and enhanced the ability of hMOF to bind to the Sirt6 promoter in OC cells, and repressed the proliferation of SKOV3 cells and promoted the apoptosis of SKOV3 cells via up-regulating Sirt6. Moreover, it was found that miR-572 negatively regulated hMOF luciferase activity. After the transfection of miR-572 inhibitor into SKOV3 cells, the cell proliferation was significantly repressed, while this repression was reversed after the transfection of shRNA-hMOF. Besides, the overexpression of hMOF could significantly inhibit the growth of tumors. Overall, our findings uncovered a novel regulatory pattern of hMOF in OC progression and provided new insights for relieving OC.