MilliporeSigma
  • Video-based assessment of drug-induced effects on contractile motion properties using human induced pluripotent stem cell-derived cardiomyocytes.
Video-based assessment of drug-induced effects on contractile motion properties using human induced pluripotent stem cell-derived cardiomyocytes.
Journal of pharmacological and toxicological methods (2020-07-04)
Yoshiko Okai, Keiko Matsune, Kazunori Yamanaka, Toshikatsu Matsui, Emily Pfeiffer Kaushik, Kosuke Harada, Hiroshi Kohara, Akimitsu Miyawaki, Harushige Ozaki, Matthew Wagoner, Tadahiro Shinozawa
ABSTRACT
Drug-induced inotropic change is a risk factor in drug development; thus, de-risking is desired in the early stages of drug development. Unlike proarrhythmic risk assessment using human induced pluripotent stem cell-derived cardiomyocytes (hiPSC-CMs), few in vitro models were validated to predict cardiac contractility. Motion field imaging (MFI), a high-resolution block matching-based optical flow technique, was expected to possess high quantitative predictivity in the detection of contraction speed. We aimed to establish an in vitro model to assess drug-induced contractile changes using hiPSC-CMs and MFI. MFI was designed to noninvasively characterize cardiomyocyte contractile behavior by analyzing light microscope video images, and maximum contraction speed (MCS) was used as the index of contractility. Using MFI, 9 inactive compounds, 10 negative inotropes, and 10 positive inotropes were tested. Two negative chronotropes, ZD7288 and ivabradine, were also tested. To determine the sensitivity and specificity of the assay, the minimum effective concentration of the MCS was compared with the human effective total therapeutic concentration for 28 compounds in clinical use. For 8 negative and 8 positive inotropes, the effects observed in in vivo and clinical studies were detected in MFI assay. MFI assay showed negative chronotropic changes without inotropic changes. MFI assay presented sufficient specificity (83%) and sensitivity (88%), and RNA-sequence analysis provided an insight into the relationship between occurrence of the false compounds and target gene expression. We demonstrated the utility of MFI assay using hiPSC-CMs to assess drug-induced contractile function. These results will facilitate the de-risking of compounds during early drug development.
Sigma-Aldrich
ZD7288 hydrate, ≥98% (HPLC)
Sigma-Aldrich
Zimelidine dihydrochloride, solid
Sigma-Aldrich
Ivabradine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Enoximone, solid