MilliporeSigma
  • Vemurafenib may overcome TNF-related apoptosis-inducing ligand (TRAIL) resistance in anaplastic thyroid cancer cells.
Vemurafenib may overcome TNF-related apoptosis-inducing ligand (TRAIL) resistance in anaplastic thyroid cancer cells.
Endocrine (2019-08-05)
Tania Pilli, Silvia Cantara, Carlotta Marzocchi, Furio Pacini, Bellur S Prabhakar, Maria Grazia Castagna
ABSTRACT
Anaplastic thyroid cancer (ATC) is rare but with poor prognosis. TRAIL can selectively induce apoptosis in cancer cells; however, resistance is quite common. Aim of our study was to evaluate TRAIL-induced apoptosis in ATC-derived cell lines, in vitro and in vivo, and the effect of combination with tyrosine kinase inhibitors (TKIs) selective for BRAF (vemurafenib) or Akt (MK-2206). Four ATC-derived cell lines were used: C643, CAL62, HTh7, with activating mutation of RAS and copy gain of PI3K (HTh7) and, 8505C with activating mutation of BRAF. Cells were treated with TRAIL alone or in combination with vemurafenib or MK-2206. The pro-apoptotic effect of TRAIL alone or combined with TKIs was, also, evaluated in two mouse xenograft models (HTh7 and 8505C). C643, CAL62, and HTh7 cells were sensitive to TRAIL-induced apoptosis, whereas 8505C cells were resistant. Both in vitro and in vivo vemurafenib was able to increase the TRAIL-induced apoptosis in 8505C cells causing a slower tumor growth in 8505C xenograft compared to placebo, while MK-2206 did not have any additive effect on TRAIL treatment in HTh7 model. TRAIL is a promising therapeutic agent in ATC and in case of resistance vemurafenib may be a valid complementary therapy.