MilliporeSigma
  • Conventional and unconventional interactions of the transcription factor FOXL2 uncovered by a proteome-wide analysis.
Conventional and unconventional interactions of the transcription factor FOXL2 uncovered by a proteome-wide analysis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-01-10)
May Penrad-Mobayed, Caroline Perrin, Laetitia Herman, Anne-Laure Todeschini, Fabienne Nigon, Bertrand Cosson, Sandrine Caburet, Reiner A Veitia
ABSTRACT
Beyond the study of its transcriptional target genes, the identification of the various interactors of a transcription factor (TF) is crucial to understand its diverse cellular roles. We focused on FOXL2, a winged-helix forkhead TF important for ovarian development and maintenance. FOXL2 has been implicated in diverse cellular processes, including apoptosis, the control of cell cycle or the regulation of steroid hormone synthesis. To reliably identify partners of endogenous FOXL2, we performed a proteome-wide analysis using co-immunoprecipitation in the murine granulosa cell-derived AT29c and the pituitary-derived alpha-T3 cell lines, using three antibodies targeting different parts of the protein. Following a stringent selection of mass spectrometry data on the basis of identification reliability and protein enrichment, we identified a core set of 255 partners common to both cell lines. Their analysis showed that we could co-precipitate several complexes involved in mRNA processing, chromatin remodeling and DNA replication and repair. We further validated (direct and/or indirect) interactions with selected partners, suggesting an unexpected role for FOXL2 in those processes. Overall, this comprehensive analysis of the endogenous FOXL2 interactome sheds light on its numerous and diverse interactors and unconventional cellular roles.
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.