MilliporeSigma
  • The HERC1 ubiquitin ligase regulates presynaptic membrane dynamics of central synapses.
The HERC1 ubiquitin ligase regulates presynaptic membrane dynamics of central synapses.
Scientific reports (2020-07-23)
Mª Angeles Montes-Fernández, Eva Mª Pérez-Villegas, Francesc R Garcia-Gonzalo, Leonardo Pedrazza, Jose Luis Rosa, Guillermo Alvarez de Toledo, José A Armengol
ABSTRACT
HERC1 is a ubiquitin ligase protein, which, when mutated, induces several malformations and intellectual disability in humans. The animal model of HERC1 mutation is the mouse tambaleante characterized by: (1) overproduction of the protein; (2) cerebellar Purkinje cells death by autophagy; (3) dysregulation of autophagy in spinal cord motor neurons, and CA3 and neocortical pyramidal neurons; (4) impairment of associative learning, linked to altered spinogenesis and absence of LTP in the lateral amygdala; and, (5) motor impairment due to delayed action potential transmission, decrease synaptic transmission efficiency and altered myelination in the peripheral nervous system. To investigate the putative role of HERC1 in the presynaptic dynamics we have performed a series of experiments in cultured tambaleante hippocampal neurons by using transmission electron microscopy, FM1-43 destaining and immunocytochemistry. Our results show: (1) a decrease in the number of synaptic vesicles; (2) reduced active zones; (3) less clathrin immunoreactivity and less presynaptic endings over the hippocampal main dendritic trees; which contrast with (4) a greater number of endosomes and autophagosomes in the presynaptic endings of the tambaleante neurons relative to control ones. Altogether these results show an important role of HERC1 in the regulation of presynaptic membrane dynamics.
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
SynaptoGreen C4, ≥95% (HPLC), solid