MilliporeSigma
  • Selective serotonin reuptake inhibitors (SSRI) affect murine bone lineage cells.
Selective serotonin reuptake inhibitors (SSRI) affect murine bone lineage cells.
Life sciences (2020-05-26)
Emily Durham, Yuhua Zhang, Amanda LaRue, Amy Bradshaw, James Cray
ABSTRACT
Data suggest pharmacological treatment of depression with selective serotonin reuptake inhibitors (SSRI) may impair bone health. Our group has previously modeled compromised craniofacial healing after treatment with sertraline, a commonly prescribed SSRI, and hypothesized potential culprits: alterations in bone cells, collagen, and/or inflammation. Here we interrogate bone lineage cell alterations due to sertraline treatment as a potential cause of the noted compromised bone healing. Murine pre-osteoblast, pre-osteoclast, osteoblast, and osteoclast cells were treated with clinically relevant concentrations of the SSRI. Studies focused on serotonin pathway targets, cell viability, apoptosis, differentiation, and the osteoblast/osteoclast feedback loop. All cells studied express neurotransmitters (e.g. serotonin transporter, SLC6A4, SSRI target) and G-protein-coupled receptors associated with the serotonin pathway. Osteoclasts presented the greatest native expression of Slc6a4 with all cell types exhibiting decreases in Slc6a4 expression after SSRI treatment. Pre-osteoclasts exhibited alteration to their differentiation pathway after treatment. Pre-osteoblasts and osteoclasts showed reduced apoptosis after treatment but showed no significant differences in functional assays. OPG mRNA and protein ratios were decreased in the osteoblast lineage. Osteoclast lineage cells treated with sertraline demonstrated diminished TRAP positive cells when pre-exposed to sertraline prior to RANKL-induced differentiation. These data suggest osteoclasts are a likely target of bone homeostasis disruption due to sertraline treatment, most potently through the osteoblast/clast feedback loop.
Sigma-Aldrich
Fast Red Violet LB Salt, Dye content ≥90 %
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)