MilliporeSigma
  • Wdr47 Controls Neuronal Polarization through the Camsap Family Microtubule Minus-End-Binding Proteins.
Wdr47 Controls Neuronal Polarization through the Camsap Family Microtubule Minus-End-Binding Proteins.
Cell reports (2020-04-23)
Yawen Chen, Jianqun Zheng, Xiaowei Li, Lei Zhu, Zhifeng Shao, Xiumin Yan, Xueliang Zhu
ABSTRACT
Neurons require proper polarization for precise positioning and axon-dendrite formation. Their intrinsic regulators and underlying mechanisms are poorly understood. Here, we show that Wdr47 is a key polarization regulator. Wdr47-deficient newborn mice die of suffocation due to central nervous system defects including axonal tracts agenesis and slowed radial migration. Wdr47 deficiency represses the multipolar-bipolar transition of cortical neurons, reduces neurite tip-directed microtubule dynamics, and causes multi-axon formation. Overexpression of Wdr47 in wild-type neurons inhibits axon specification and neutralizes Taxol-induced neurite overgrowth and axon overproduction. Wdr47 interacts with the Camsap family of microtubule minus-end-binding proteins; associates with microtubules through Camsap3, whose gene disruption also causes multi-axons; and promotes Camsap3 accumulation in neurites of unpolarized neurons. Furthermore, Camsap overexpression rescues the polarization defects of Wdr47-deficient neurons. Our results indicate that Wdr47 facilitates proper neurite remodeling through Camsaps to fine-tune regional microtubule dynamics and organization during early stages of neuronal polarization.
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Histodenz, nonionic density gradient medium
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Tubulin Antibody, Detyrosinated, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Anti-Microtubule-Associated Protein 2 (MAP2) Antibody, Chemicon®, from rabbit
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder