MilliporeSigma
  • POMK regulates dystroglycan function via LARGE1-mediated elongation of matriglycan.
POMK regulates dystroglycan function via LARGE1-mediated elongation of matriglycan.
eLife (2020-09-26)
Ameya S Walimbe, Hidehiko Okuma, Soumya Joseph, Tiandi Yang, Takahiro Yonekawa, Jeffrey M Hord, David Venzke, Mary E Anderson, Silvia Torelli, Adnan Manzur, Megan Devereaux, Marco Cuellar, Sally Prouty, Saul Ocampo Landa, Liping Yu, Junyu Xiao, Jack E Dixon, Francesco Muntoni, Kevin P Campbell
ABSTRACT
Matriglycan [-GlcA-β1,3-Xyl-α1,3-]n serves as a scaffold in many tissues for extracellular matrix proteins containing laminin-G domains including laminin, agrin, and perlecan. Like-acetyl-glucosaminyltransferase 1 (LARGE1) synthesizes and extends matriglycan on α-dystroglycan (α-DG) during skeletal muscle differentiation and regeneration; however, the mechanisms which regulate matriglycan elongation are unknown. Here, we show that Protein O-Mannose Kinase (POMK), which phosphorylates mannose of core M3 (GalNAc-β1,3-GlcNAc-β1,4-Man) preceding matriglycan synthesis, is required for LARGE1-mediated generation of full-length matriglycan on α-DG (~150 kDa). In the absence of Pomk gene expression in mouse skeletal muscle, LARGE1 synthesizes a very short matriglycan resulting in a ~ 90 kDa α-DG which binds laminin but cannot prevent eccentric contraction-induced force loss or muscle pathology. Solution NMR spectroscopy studies demonstrate that LARGE1 directly interacts with core M3 and binds preferentially to the phosphorylated form. Collectively, our study demonstrates that phosphorylation of core M3 by POMK enables LARGE1 to elongate matriglycan on α-DG, thereby preventing muscular dystrophy.
Sigma-Aldrich
Calpain Inhibitor I, ≥97% (TLC), powder
Sigma-Aldrich
Cell Dissociation Solution Non-enzymatic 1x, Prepared in phosphate buffered saline without calcium and magnesium, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Leupeptin, Hemisulfate, Microbial, Leupeptin hemisulfate, CAS 103476-89-7, is a reversible inhibitor of trypsin-like proteases and cysteine proteases
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Leucoberbelin Blue I, Dye content 65 %