Merck
  • SPT6-driven error-free DNA repair safeguards genomic stability of glioblastoma cancer stem-like cells.
SPT6-driven error-free DNA repair safeguards genomic stability of glioblastoma cancer stem-like cells.
Nature communications (2020-09-20)
Elisabeth Anne Adanma Obara, Diana Aguilar-Morante, Rikke Darling Rasmussen, Alex Frias, Kristoffer Vitting-Serup, Yi Chieh Lim, Kirstine Juul Elbæk, Henriette Pedersen, Lina Vardouli, Kamilla Ellermann Jensen, Jane Skjoth-Rasmussen, Jannick Brennum, Lucie Tuckova, Robert Strauss, Christoffel Dinant, Jiri Bartek, Petra Hamerlik
ABSTRACT
Glioblastoma cancer-stem like cells (GSCs) display marked resistance to ionizing radiation (IR), a standard of care for glioblastoma patients. Mechanisms underpinning radio-resistance of GSCs remain largely unknown. Chromatin state and the accessibility of DNA lesions to DNA repair machineries are crucial for the maintenance of genomic stability. Understanding the functional impact of chromatin remodeling on DNA repair in GSCs may lay the foundation for advancing the efficacy of radio-sensitizing therapies. Here, we present the results of a high-content siRNA microscopy screen, revealing the transcriptional elongation factor SPT6 to be critical for the genomic stability and self-renewal of GSCs. Mechanistically, SPT6 transcriptionally up-regulates BRCA1 and thereby drives an error-free DNA repair in GSCs. SPT6 loss impairs the self-renewal, genomic stability and tumor initiating capacity of GSCs. Collectively, our results provide mechanistic insights into how SPT6 regulates DNA repair and identify SPT6 as a putative therapeutic target in glioblastoma.
Sigma-Aldrich
5-Ethynyl uridine, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mg
Available to ship on 07 April 2025
Details...
$19.80
5 mg
Available to ship on 07 April 2025
Details...
$99.60
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission, certified by the BSC
Pricing and availability is not currently available.
Sigma-Aldrich
Agarose, low gelling temperature, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Available to ship on 07 April 2025
Details...
$264.00
100 g
Please contact Customer Service for Availability
$1,610.00
250 g
Please contact Customer Service for Availability
$3,620.00