MilliporeSigma
  • Polymyxin B for the treatment of multidrug-resistant pathogens: a critical review.
Polymyxin B for the treatment of multidrug-resistant pathogens: a critical review.
The Journal of antimicrobial chemotherapy (2007-09-20)
Alexandre Prehn Zavascki, Luciano Zubaran Goldani, Jian Li, Roger L Nation
ABSTRACT
Polymyxins have re-emerged in clinical practice owing to the dry antibiotic development pipeline and worldwide increasing prevalence of nosocomial infections caused by multidrug-resistant (MDR) Gram-negative bacteria. Polymyxin B and colistin (polymyxin E) have been ultimately considered as the last-resort treatment of such infections. Microbiological, pharmacokinetic, pharmacodynamic and clinical data available for polymyxin B are reviewed in this paper. Polymyxin B has rapid in vitro bactericidal activity against major MDR Gram-negative bacteria, such as Pseudomonas aeruginosa, Acinetobacter baumannii and Klebsiella pneumoniae. Acquired resistance to this agent is still rare among these pathogens. However, optimized dosage regimens are not known yet. Good clinical outcomes have been observed in the majority of the patients treated with intravenous polymyxin B in recent studies. However, these studies failed to provide definitive conclusions due to limitations of study design and additional clinical trials are required. Although combination therapy may be an attractive option based on some currently available in vitro data, clinical data supporting such recommendations are lacking. Since polymyxins will be increasingly used for the treatment of infections caused by MDR bacteria, clinical pharmacokinetic, pharmacodynamic and toxicodynamic studies underpinning the optimal use of these drugs are urgently required.
Sigma-Aldrich
Rhodamine B labeled Polymyxin B Ready Made Solution, For fluorescent microbial imaging, 1.1 mg/mL in water
Sigma-Aldrich
Dansyl labeled polymyxin B Ready Made Solution, for fluorescent microbial imaging, 1.5 mg/mL in H2O
Sigma-Aldrich
Polymyxin B sulfate salt