MilliporeSigma
  • Structural and functional insight into the effect of AFF4 dimerization on activation of HIV-1 proviral transcription.
Structural and functional insight into the effect of AFF4 dimerization on activation of HIV-1 proviral transcription.
Cell discovery (2020-03-05)
Dan Tang, Chunjing Chen, Ga Liao, Jiaming Liu, Banghua Liao, QingQing Huang, Qianqian Chen, Jiahui Zhao, Hui Jiang, Jinsong Duan, Jin Huang, Kunjie Wang, Jiawei Wang, Cuiyan Zhou, Wendan Chu, Wenqi Li, Bo Sun, Zhonghan Li, Lunzhi Dai, Xianghui Fu, Wei Cheng, Yuhua Xue, Shiqian Qi
ABSTRACT
Super elongation complex (SEC) is a positive regulator of RNA polymerase II, which is required for HIV-1 proviral transcription. AFF1/4 is the scaffold protein that recruits other components of SEC and forms dimer depending on its THD domain (TPRL with Handle Region Dimerization Domain). Here we report the crystal structure of the human AFF4-THD at the resolution of 2.4 Å. The α4, α5, and α6 of one AFF4-THD mediate the formation of a dimer and pack tightly against the equivalent part of the second molecule in the dimer of AFF-THD. Mutagenesis analysis revealed that single mutations of either Phe1014 or Tyr1096 of AFF4 to alanine impair the formation of the AFF4 dimer. In addition, transactivation assay also indicated that Phe1014 and Tyr1096 of AFF4 are critical to the transactivation activity of AFF4. Interestingly, the corresponding residues Phe1063 and Tyr1145 in AFF1 have an effect on the transactivation of HIV-1 provirus. However, such mutations of AFF1/4 have no effect on the interaction of AFF1/4 with other subunits of the SEC. Together, our data demonstrated that the dimerization of AFF1/4 is essential to transactivation of HIV-1 provirus.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid