MilliporeSigma
  • Development of a direct competitive enzyme-linked immunosorbent assay for determination of the fungicide mepanipyrim and its metabolite.
Development of a direct competitive enzyme-linked immunosorbent assay for determination of the fungicide mepanipyrim and its metabolite.
Journal of pesticide science (2019-09-19)
Shiro Miyake, Yuki Hirakawa, Tomomi Yamasaki, Eiki Watanabe, Ayako Harada, Kayo Adachi, Seiji Iwasa, Hiroshi Narita
ABSTRACT
A direct competitive enzyme-linked immunosorbent assay (dc-ELISA) was developed for determination of anilinopyrimidine fungicide mepanipyrim in vegetables. Two derivatives of mepanipyrim and mepanipyrim propanol type metabolite which carried carboxy acid were synthesized and conjugated with keyhole limpet hemocyanin. BALB/c mice were immunized to prepare anti-mepanipyrim monoclonal antibodies (MoAbs) by obtained conjugates. The dc-ELISAs based on the prepared MoAbs, MPP107 and MPP204, showed working ranges between 0.12 and 1.8 ng/mL with mepanipyrim for MPP107, 0.12 and 2.4 ng/mL with mepanipyrim for MPP204, and 0.2 ng/mL and 5.7 ng/mL with the mepanipyrim propanol type for MPP204. The dc-ELISAs showed the sufficient sensitivity to determine the mepanipyrim residues for the MRLs of 1-15 mg/kg among the majority of vegetables and fruits in Japan. Recovery and/or correlation results from HPLC suggested that the dc-ELISAs would be applicable to the residue analysis of mepanipyrim and its propanol type in vegetables.
Product Description
Supelco
Cyprodinil, PESTANAL®, analytical standard
Supelco
Pyrimethanil, PESTANAL®, analytical standard
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
Sulfuric acid solution, 0.5 M