MilliporeSigma
  • Huh-7 or HepG2 cells: which is the better model for studying human apolipoprotein-B100 assembly and secretion?
Huh-7 or HepG2 cells: which is the better model for studying human apolipoprotein-B100 assembly and secretion?
Journal of lipid research (2010-10-20)
Steven J R Meex, Ursula Andreo, Janet D Sparks, Edward A Fisher
ABSTRACT
Apolipoprotein-B100 (apoB100) is the essential protein for the assembly and secretion of very low density lipoproteins (VLDL) from liver. The hepatoma HepG2 cell line has been the cell line of choice for the study of synthesis and secretion of human apoB-100. Despite the general use of HepG2 cells to study apoB100 metabolism, they secrete relatively dense, lipid-poor particles compared with VLDL secreted in vivo. Recently, Huh-7 cells were adopted as an alternative model to HepG2 cells, with the implicit assumption that Huh-7 cells were superior in some respects of lipoprotein metabolism, including VLDL secretion. In this study we addressed the hypothesis that the spectrum of apoB100 lipoprotein particles secreted by Huh-7 cells more closely resembles the native state in human liver. We find that Huh-7 cells resemble HepG2 cells in the effects of exogenous lipids, microsomal triglyceride transfer protein (MTP)-inhibition, and proteasome inhibitors of apoB100 secretion, recovery, and degradation. In contrast to HepG2 cells, however, MEK-ERK inhibition does not correct the defect in VLDL secretion. Huh-7 cells do not appear to offer any advantages over HepG2 cells as a general model of human apoB100-lipoprotein metabolism.
Product Description
Huh-7D 12 Cell Line human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. 01042712, human liver, Epithelial
Sigma-Aldrich
Anti-Apolipoprotein B Goat pAb, liquid, Calbiochem®