MilliporeSigma
  • Converting bleomycin into a prodrug that undergoes spontaneous reactivation under physiological conditions.
Converting bleomycin into a prodrug that undergoes spontaneous reactivation under physiological conditions.
Toxicology and applied pharmacology (2019-10-28)
Itzik Cooper, Dana Atrakchi, Michael D Walker, Amnon Horovitz, Mati Fridkin, Yoram Shechter
ABSTRACT
Bleomycin is an anticancer antibiotic effective against a range of human malignancies. Yet its usefulness is limited by serious side effects. In this study, we converted bleomycin into a prodrug by covalently linking 2-sulfo, 9 fluorenylmethoxycarbonyl (FMS) to the primary amino side chain of bleomycin. FMS-bleomycin lost its efficacy to bind transition metal ions and therefore was converted into an inactive derivative. Upon incubation in vitro under physiological conditions, the FMS-moiety undergoes spontaneous hydrolysis, generating native bleomycin possessing full anti-bacterial potency. FMS hydrolysis and reactivation takes place with a t1/2 value of 17 ± 1 h. In silico simulation predicts a narrow therapeutic window in human patients of seven hours, starting 40 min after administration. In mice, close agreement was obtained between the experimental and the simulated pharmacokinetic profiles for FMS-bleomycin. FMS-bleomycin is thus shown to be a classical prodrug: it is inactive at the time of administration and the non-modified (active) bleomycin is released with a desirable pharmacokinetic profile following administration, suggesting it may have therapeutic value in the clinic.
Sigma-Aldrich
Dialysis tubing, benzoylated, Avg. flat width 32 mm (1.27 in.)