MilliporeSigma
  • Fractionated Antioxidant and Anti-inflammatory Kernel Oil from Torreya fargesii.
Fractionated Antioxidant and Anti-inflammatory Kernel Oil from Torreya fargesii.
Molecules (Basel, Switzerland) (2019-09-25)
Xianrong Zhou, Jin Shang, Mingyi Qin, Jianhua Wang, Bo Jiang, Hui Yang, Yan Zhang
ABSTRACT
Polymethylene-interrupted polyunsaturated fatty acids (PMI-PUFAs) are emerging functional lipids with proven antioxidant and anti-inflammatory effects. In this study, a typical PMI-PUFA, sciadonic acid (C20:3, 5c 11c 14c), was enriched in the kernel oil of Torreya fargesii (T. fargesii) by fractionation. Fractionated kernel oil of T. fargesii (containing 25% sciadonic acid) showed equal stability and similar radical scavenging ability compared with the non-fractionated oil. In anti-inflammatory tests, fractionated kernel oil was shown to inhibit the activity of phosphodiesterase (PDE-5, efficiency 80% at 133.7 μg/mL) and lipoxygenase-5 (LOX-5, efficiency 65% at 66.7 μg/mL) more effectively than the non-fractionated oil. This shows that increasing the amount of sciadonic acid can enhance the anti-inflammatory effect of the kernel oil. This research also indicates that fractionation is a feasible way to obtain sciadonic acid-rich functional oil with potential pharmacological effects.
Sigma-Aldrich
Lipoxidase from Glycine max (soybean), Type V, ammonium sulfate suspension, 500,000-1,000,000 units/mg protein
Supelco
Squalene, analytical standard
Supelco
F.A.M.E. Mix, C18:0 - C20:0, certified reference material, pkg of 100 mg (Neat)