MilliporeSigma
  • Deep Conservation of cis-Element Variants Regulating Plant Hormonal Responses.
Deep Conservation of cis-Element Variants Regulating Plant Hormonal Responses.
The Plant cell (2019-08-31)
Michal Lieberman-Lazarovich, Chen Yahav, Alon Israeli, Idan Efroni
ABSTRACT
Phytohormones regulate many aspects of plant life by activating transcription factors (TFs) that bind sequence-specific response elements (REs) in regulatory regions of target genes. Despite their short length, REs are degenerate, with a core of just 3 to 4 bp. This degeneracy is paradoxical, as it reduces specificity and REs are extremely common in the genome. To study whether RE degeneracy might serve a biological function, we developed an algorithm for the detection of regulatory sequence conservation and applied it to phytohormone REs in 45 angiosperms. Surprisingly, we found that specific RE variants are highly conserved in core hormone response genes. Experimental evidence showed that specific variants act to regulate the magnitude and spatial profile of hormonal response in Arabidopsis (Arabidopsis thaliana) and tomato (Solanum lycopersicum). Our results suggest that hormone-regulated TFs bind a spectrum of REs, each coding for a distinct transcriptional response profile. Our approach has implications for precise genome editing and for rational promoter design.
Sigma-Aldrich
Picloram, BioReagent, suitable for plant cell culture
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, ≥95%, crystalline