MilliporeSigma
  • Fabricating Fibers of a Porous-Polystyrene Shell and Particle-Loaded Core.
Fabricating Fibers of a Porous-Polystyrene Shell and Particle-Loaded Core.
Molecules (Basel, Switzerland) (2019-11-17)
Dharneedar Ravichandran, Weiheng Xu, Rahul Franklin, Namrata Kanth, Sayli Jambhulkar, Sumedh Shukla, Kenan Song
ABSTRACT
Polystyrene (PS) polymers have broad applications in protective packaging for food shipping, containers, lids, bottles, trays, tumblers, disposable cutlery and the making of models. Currently, most PS products, such as foams, are not accepted for recycling due to a low density in the porous structure. This poses a challenge for logistics as well as creating a lack of incentive to invest in high-value products. This study, however, demonstrated the use of a dry-jet wet-spinning technique to manufacture continuous PS fibers enabled by an in-house designed and developed spinning apparatus. The manufactured fibers showed porosity in the shell and the capability to load particles in their core, a structure with high potential use in environmentally relevant applications such as water treatment or CO2 collections. A two-phase liquid-state microstructure was first achieved via a co-axial spinneret. Following coagulation procedures and heat treatment, phase-separation-based selective dissolution successfully generated the porous-shell/particle-core fibers. The pore size and density were controlled by the porogen (i.e., PEG) concentrations and examined using scanning electron microscopy (SEM). Fiber formation dynamics were studied via rheology tests and gelation measurements. The shell components were characterized by tensile tests, thermogravimetric analysis, and differential scanning calorimetry for mechanical durability and thermal stability analyses.
Sigma-Aldrich
Polystyrene, average Mw ~350,000, average Mn ~170,000
Sigma-Aldrich
Polystyrene, average Mw ~192,000
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Sigma-Aldrich
Poly(ethylene glycol), average Mn 10,000, flakes