MilliporeSigma
  • IL‑1β increases the expression of inflammatory factors in synovial fluid‑derived fibroblast‑like synoviocytes via activation of the NF‑κB‑mediated ERK‑STAT1 signaling pathway.
IL‑1β increases the expression of inflammatory factors in synovial fluid‑derived fibroblast‑like synoviocytes via activation of the NF‑κB‑mediated ERK‑STAT1 signaling pathway.
Molecular medicine reports (2019-10-23)
Jie Yang, Junhu Wang, Xiaojun Liang, Hongmou Zhao, Jun Lu, Qiang Ma, Bingfei Jing, Feng Tian
ABSTRACT
Interleukin (IL)‑1β serves a crucial role in the progression of rheumatoid arthritis. Previous studies have indicated that the ERK/STAT1 signaling pathway may be involved in the inflammatory response in synovial fluid‑derived fibroblast‑like synoviocytes (sfd‑FLSs). However, the molecular mechanisms underlying the pathological effects of the inflammatory factors induced by IL‑1β in sfd‑FLSs remain unclear. The aim of the present study was to investigate the IL‑1β‑mediated signaling pathways involved in the expression of inflammatory factors in sfd‑FLSs and in a rat model of rheumatoid arthritis. Reverse transcription‑quantitative PCR, western blotting, and immunohistochemistry were used to analyze the role of IL‑1β in the rat model of rheumatoid arthritis. The results suggested that IL‑1β administration exacerbated rheumatoid arthritis, bone injury and increased the expression levels of inflammatory factors, such as IL‑17 and tumor necrosis factor α (TNF‑α), whereas treatment with anti‑IL‑1β exhibited opposite effects. In vitro experiments in sfd‑FLSs further suggested that treatment with IL‑1β influenced the expression levels of various inflammatory factors. In specific, IL‑1β increased the expression of IL‑17 and TNF‑α, and decreased the expression of IL‑6 and IL‑10 in sfd‑FLSs. Additionally, treatment with IL‑1β increased the mRNA expression and protein phosphorylation of NF‑κB, ERK and STAT1 in sfd‑FLSs. Treatment with anti‑IL‑1β exhibited opposite effects on the expression levels of inflammatory factors and suppressed the NF‑κB‑mediated ERK‑STAT1 signaling pathway activation in sfd‑FLSs. Finally, treatment with a NF‑κB inhibitor suppressed the effects of IL‑1β, and NF‑κB overexpression reversed the effects of anti‑IL‑1β on the expression levels of IL‑17, TNF‑α, NF‑κB, ERK and STAT1. In conclusion, the present results demonstrated that treatment with IL‑1β increased the expression levels of inflammatory factors in sfd‑FLSs via the regulation of the NF‑κB‑mediated ERK/STAT1 signaling pathway in a rat model of rheumatoid arthritis. Therefore, the NF‑κB/ERK/STAT1 signaling pathway may represent a potential target for the development of novel treatments for rheumatoid arthritis.
Sigma-Aldrich
NF-kB Activation Inhibitor IV, The NF-kB Activation Inhibitor IV, also referenced under CAS 139141-12-1, controls the biological activity of NF-kB. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.
Sigma-Aldrich
Anti-IL-17 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
IL 17 A/F from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE)