- Gankyrin promotes osteosarcoma tumorigenesis by forming a positive feedback loop with YAP.
Gankyrin promotes osteosarcoma tumorigenesis by forming a positive feedback loop with YAP.
Although gankyrin has been identified as a vital regulator of tumorigenesis, its role and regulatory mechanism in osteosarcoma (OS) remain unclear. QRT-PCR, western blot and IHC staining were conducted to detect the expression of gankyrin in OS. Pearson's χ² test was adopted to examine the associations between gankyrin expression and clinicopathologic characteristics. Kaplan-Meier method was used to investigate the relationship between gankyrin expression and overall survival of patients with OS. Next, a series of in vitro and in vivo assays were performed to determine the positive feedback loop between gankyrin and YAP in OS. We first reported that gankyrin is upregulated in human OS specimens and cell lines and predicts OS progression and poor prognosis. Furthermore, we demonstrated that gankyrin protects miR-200a-mediated yes-associated protein (YAP) downregulation through p53 and establishes a positive feedback loop to regulate YAP signaling in U2OS and MG63 cells. Intriguingly, gankyrin interacts with YAP to promote OS cell growth in vitro. In addition, our results showed that gankyrin promotes OS tumor growth and regulates YAP levels in vivo. Notably, we also observed a positive correlation between gankyrin and YAP expression in human OS tissues, and co-upregulation of gankyrin and YAP indicated a poor prognosis. Our results identify that gankyrin acts as an oncogene in OS by forming a positive feedback loop with YAP, and disrupting the gankyrin-YAP regulation may be beneficial for controlling OS tumorigenesis.