MilliporeSigma
Clec10a regulates mite-induced dermatitis.
Science immunology (2019-12-08)
Kazumasa Kanemaru, Emiko Noguchi, Satoko Tahara-Hanaoka, Seiya Mizuno, Hiroaki Tateno, Kaori Denda-Nagai, Tatsuro Irimura, Hiroshi Matsuda, Fumihiro Sugiyama, Satoru Takahashi, Kazuko Shibuya, Akira Shibuya
ABSTRACT
House dust mite (HDM) is a major allergen that causes allergic diseases such as atopic dermatitis. However, the regulatory mechanisms of HDM-induced immune responses are incompletely understood. NC/Nga mice are an inbred strain that is more susceptible to HDM and develops more severe dermatitis than other strains. Using whole-exome sequencing, we found that NC/Nga mice carry a stop-gain mutation in Clec10a, which encodes a C-type lectin receptor, Clec10a (MGL1/CD301a). The repair of this gene mutation using the CRISPR-Cas9 system ameliorated HDM-induced dermatitis, indicating that the Clec10a mutation is responsible for hypersensitivity to HDM in NC/Nga mice. Similarly, Clec10a-/- mice on the C57BL/6J background showed exacerbated HDM-induced dermatitis. Clec10a expressed on skin macrophages inhibits HDM-induced Toll-like receptor 4 (TLR4)-mediated inflammatory cytokine production through the inhibitory immunoreceptor tyrosine activating motif in its cytoplasmic portion. We identified asialoglycoprotein receptor 1 (Asgr1) as a functional homolog of mouse Clec10a in humans. Moreover, we found that a mucin-like molecule in HDM is a ligand for mouse Clec10a and human Asgr1. Skin application of the ligand ameliorated a TLR4 ligand-induced dermatitis in mice. Our findings suggest that Clec10a in mice and Asgr1 in humans play an important role in skin homeostasis against inflammation associated with HDM-induced dermatitis.
Sigma-Aldrich
MISSION® esiRNA, targeting human ASGR1
Sigma-Aldrich
Syk Inhibitor IV, BAY 61-3606, The Syk Inhibitor IV, BAY 61-3606, also referenced under CAS 732938-37-8, controls the biological activity of Syk. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Octoclothepin maleate salt, solid